Leupeptin hemisulphate
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Powder
MDL No.: MFCD00037012
CAS No.: 103476-89-7
Molecular Formula: C20H38N6O4.0.5H2SO4
Formula Weight: 475.59
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water at 50mg/ml
MDL No.: MFCD00037012
Reversible protease inhibitor which inhibits cathepsin B, calpain and trypsin
Related Information
Customers Who Bought This Also Bought