Insulin B
Catalog #: CAAAJ61170-MB
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:25 mg
- MDL No.:MFCD00135750
- Protein/peptide name:Insulin B
- Cat. no.:CAAAJ61170-MB
- Supplier No.:J61170-MB
Specifications
About this item
Powder
CAS No.: 34367-73-2
Molecular Formula: C26H35N7O5
Formula Weight: 525.61
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00135750
Customers Who Bought This Also Bought