Transformation Reagent, TB Solution-II, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Used for the transoformation of competent cells.
Contains 250 mM potassium chloride, 15 mM calcium chloride, 10mM HEPES and 55 mM manganese chloride, pH 6.5.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.