Order Entry
ContactUsLinkComponent
Medium pH Meter Supplies Organizer, PVC, TrippNT
Medium pH Meter Supplies Organizer, PVC, TrippNT
Catalog #: 11376-300
Supplier:  TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Medium pH Meter Supplies Organizer
Medium pH Meter Supplies Organizer, PVC, TrippNT
Catalog #: 11376-300
Supplier:  TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Medium pH Meter Supplies Organizer
  • Exterior W×D×H:
    61 x 15 x 30 cm (24 x 6 x 12")
  • Weight:
    2 kg (5 lbs.)
  • Supplier No.:
    50039

Specifications

About this item

White PVC medium pH meter supplies organizer stores and protects pH meter and other lab supplies giving your lab a clean appearance.

  • Two-shelf design has a one inch safety lip that keeps supplies secure
  • Made of white, durable PVC material

It also has a built in Kimwipe wiper dispenser and a 6 hole tool holder for spatulas, thermometers or probes.

Customers Who Bought This Also Bought