Regulator, Cylinder, Diaphragm, Single Stage, Brass, Rotarex
Supplier: Ceodeux Inc.
Certificates
About this item
Designed for application as a cylinder regulator
Pressure control with Balanced-Valve Technology: the effect of inlet pressure fluctuations on outlet pressure are minimized. The BV-technology enables the delivery of a very stable outlet pressure and flow even with high flow. BV Technology also increases the useful lifetime of the regulator and reduces ownership cost.
Ideally suited for high purity gases and high-pressure applications requiring high flow and precise outlet pressure, such as for laser applications. Used also in nuclear research department where the precision of the outlet pressure and high flow are essential.
Ordering information: Additional outlet pressures and flow rates available; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought