To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Thymidine Phosphorylase
- Clonality:Monoclonal
- Clone:P-GF.44C
- Gene ID:1890
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:TYMP
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 μl
- Western blot:Yes
- Antigen synonyms:TP|MNGIE|PDECGF|ECGF1|hPD-ECGF|MEDPS1|ECGF|MTDPS1
- Modification:Thymidine phosphorylase
- Storage buffer:10mM PBS, pH7.4 containing gelatin and 0.09% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:2 mg/ml
- Immunogen:Recombinant full length human thymidine phosphorylase (TP / PD-ECGF) protein.
- Tested applications:IHC-P, IHC-Fr, WB
- Purification:Protein G purified
- Cat. no.:89351-900
- Supplier No.:GTX23151
Specifications
About this item
Mouse monoclonal [P - GF.44C] to Thymidine Phosphorylase
Western Blotting: assay dependent. Immunohistochemistry (formalin fixed paraffin embedded): use at a concentration of 1 - 2?g / ml for 30 min at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of 55 kDa, which corresponds to the predicted molecular weight of Thymidine Phosphorylase. No special pretreatment is required for immuno-histochemical staining of formalin-fixed tissues
Type: Primary
Antigen: TYMP
Clonality: Monoclonal
Clone: P-GF.44C
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat