To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Stage-specific Embryonic Antigen 3
- Clonality:Monoclonal
- Clone:MC-631
- Gene ID:93961
- Host:Rat
- Isotype:IgM
- Reactivity:Human
- Antigen symbol:SSEA3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:SSEA-3
- Storage buffer:PBS containing 0.05% BSA and 0.05% Sodium Azide
- Storage temperature:Store at -20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:4-8 cell stage mouse embryos
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Antigen affinity purification
- Cat. no.:89363-648
- Supplier No.:GTX48036
Specifications
About this item
Rat Monoclonal antibody to SSEA-3
For flow cytometry, we recommend using 0.5 ug per million cells. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: SSEA3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human