To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Serum Amyloid A
- Clonality:Monoclonal
- Clone:3H62
- Gene ID:6288
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:SAA1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:SAA2|PIG4|SAA|TP53I4
- Storage buffer:Phosphate-buffered saline, pH 7.2, containing 40% glycerol
- Molecular weight:12 kDa
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Highly purified recombinant human SAA, 12 kD
- Tested applications:ELISA, WB
- Purification:Protein G purified
- Cat. no.:89360-330
- Supplier No.:GTX17758
Specifications
About this item
The serum amyloid A (SAA) family comprises a number of differentially expressed lipoproteins, acute-phase SAA1 and SAA2, the former being a major component in plasma, and constitutive SAA's (C-SAAs). Although the liver is the primary site of synthesis of both SAA types, extrhepatic production has been reported. The in-vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed it's importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations. SAA rises earlier and more sharply than CRP.SAA enhances the binding of HDL's to macrophages and thus helps the delivery of lipid to sites of injury for use in tissue repair. It is thus thought to be an integral part of the disease process. In addition, recent experiments suggest that SAA may play a "houekeeping" role in normal human tissues.Elevated levels of SAA over time predispose secondary amyloidosis, extracellular accumulation of amyloid fibrils, derived from a circulating precursor, in various tissues and organs. The most common form of amyloidosis occurs secondary to chronic inflammatory disease, particularly rheumatoid artheritis.
For ELISA: Use at a concentration of 0.3 ?g/ml (capture coating dose). Allows detection of 1.1 ng/ml of SAA in serum/plasma or 0.6 ng/ml of SAA in medium with an assay range of 0 to 80 ng/ml. Suggested substrate: TMB. For WB: Use at a concentration of 0.1-0.5 ?g/ml to visualize 100 ng/lane of recombinant human SAA. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: SAA1
Clonality: Monoclonal
Clone: 3H62
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human