To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:PAPAIN
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Antigen symbol:PAPAIN
- Conjugation:Unconjugated
- ELISA:Yes
- Western blot:Yes
- Storage buffer:Preservative: 0.01% Sodium Azide. Constituents: PBS, 10mg/ml BSA, (IgG and protease free), pH 7.2
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Papain (Carica papaya).
- Tested applications:Immunomicroscopy
- Purification:Protein G purified
- Cat. no.:89363-254
- Supplier No.:GTX40595
Specifications
About this item
Papain is a protease derived from the papaya fruit (Carica papaya). It hydrolyses peptides with broad specificity, but shows a preference for large hydrophobic amino acids in the P2 position. It is synthesised as an inactive 40 kDa precursor containing a 107 residue N terminal pro region and a 26 residue signal peptide.
Type: Primary
Antigen: PAPAIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: