Anti-CDKN1B Mouse Monoclonal Antibody [clone: IMD-27]
Catalog #: 89363-048
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase Inhibitor 1B
- Clonality:Monoclonal
- Clone:IMD-27
- Gene ID:1027
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CDKN1B
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:CDKN4|KIP1|MEN1B|P27KIP1 |MEN4 |p27Kip1
- Storage buffer:1.2% sodium acetate or neutral PBS
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C . Avoid multiple freeze-thaw cycles.
- Immunogen:recombinant rodent p27Kip1 protein.
- Tested applications:ICC/IF, WB
- Purification:ProteinG
- Cat. no.:89363-048
- Supplier No.:GTX38504
Specifications
About this item
Mouse monoclonal antibody [IMD-27] to p27
Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Customers Who Bought This Also Bought