To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neural Cell Adhesion Molecule 1
- Clonality:Monoclonal
- Clone:UJ13A
- Gene ID:4684
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:NCAM1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:NCAM|MSK39|CD56
- Storage buffer:Phosphate buffered saline
- Storage temperature:Store at 4 °C. Long term (>6months) aliquot and store at –20 °C. Avoid repeated freeze thawing.
- Concentration:1 mg/ml
- Immunogen:16 week human foetal brain
- Tested applications:ICC/IF, WB
- Purification:Protein A purified
- Cat. no.:89360-750
- Supplier No.:GTX20036
Specifications
About this item
NCAM, as a member of the immunoglobulin superfamily of adhesion molecules is characterized by several immunoglobulin (Ig)-like domains. The extracellular part of NCAM consists of five of these Ig domains and two fibronectin type III homology regions. NCAM is encoded by a single copy gene composed of 26 exons. However, at least 20-30 distinct isoforms can be generated by alternative splicing and by posttranslational modifications, such as sialylation. During sialylation, polysialic acid (PSA) carbohydrates are attached to the extracellular part of NCAM. Through its extracellular region, NCAM mediates homophilic interactions. In addition, NCAM can also undergo heterophilic interactions by binding extracellular matrix components, such as laminin, or other cell adhesion molecules, such as integrins. NCAM is expressed on most neuroectodermal derived cell lines, tissues and neoplasm such as retinoblastoma, medulloblastoma, astrocytomas and neuroblastoma.
IF: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 120 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: UJ13A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human