To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Major Vault Protein
- Clonality:Monoclonal
- Clone:LRP-56
- Gene ID:9961
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:MVP
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:500 μl
- Format:Tissue culture supernatant
- Antigen synonyms:VAULT1|LRP
- Storage buffer:PBS with 1% BSA and 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Immunogen:2R120 cell line derived by stepwise Doxorubicin exposure of the the SW-1573 non-small cell lung carcinoma cell line.
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Tissue Culture Supernatant
- Cat. no.:89351-988
- Supplier No.:GTX23367
Specifications
About this item
Mouse monoclonal [LRP - 56] to MVP
FACS: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. IHC-Fr: Frozen tissues, cell lines, formalin-fixed/paraffin embedded tissues. Use at a dilution of 1/10 - 1/40. Pretreatment is not required. For optimal staining, the primary antibody should be incubated overnight at 4°C. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MVP
Clonality: Monoclonal
Clone: LRP-56
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human