To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) Ligand 4
- Clonality:Polyclonal
- Gene ID:20303
- Host:Goat
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Mouse
- Antigen symbol:CCL4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:SCYA4|HC21|G-26|AT744.1|LAG-1|MIP-1-beta|SCYA2|ACT2|MIP1B|MIP1B1|LAG1
- Storage buffer:Preservative: NoneConstituents: PBS. pH 7.4
- Concentration:1mg/ml
- Immunogen:Mouse recombinant macrophage inflammatory protein-1beta, expressed in E. coli, as the immunogen.
- Tested applications:ELISA, Neut, WB
- Purification:Protein G purified
- Cat. no.:89350-128
- Supplier No.:GTX10386
Specifications
About this item
Goat polyclonal to Macrophage Inflammatory Protein 1 beta
ELISA: Use at a concentration of 0.5 ?g/ml to detect <1ng/well of recombinant mouse MIP1 beta. Neut: Use at a concentration of 50 - 100 ?g/ml. WB: Use at a concentration of 1 ?g/ml to detect recombinant mouse MIP1 beta at 5ng/lane under reducing conditions. Predicted molecular weight: 11 kDa. Anti-Mouse MIP1 beta was tested for its ability to neutralize the bioactivity of recombinant, mouse MIP1 beta in an in vitro colony forming assay using hematopoietic stem cells. The ND50 of the antibody is defined as the concentration of antibody resulting in a one-half maximal inhibition of bioactivity of recombinant, mouse MIP1 beta, which is present at five times its own EC50 (the concentration of recombinant, mouse MIP1 beta producing a one-half maximal bioactivity without antibody). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CCL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse