To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 8
- Clonality:Monoclonal
- Clone:I8-S2
- Gene ID:3576
- Host:Mouse
- Isotype:IgG2b
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:IL8
- Conjugation:Unconjugated
- ELISA:Yes
- Size:250μg
- Western blot:Yes
- Antigen synonyms:MDNCF|Interleukin-8|T-cell chemotactic factor|LYNAP|NAP1|NAP-1|GCP1|Monocyte-derived neutrophil chemotactic factor|MONAP|LECT|NAF|GCP-1|CXCL8|LUCT|Neutrophil-activating protein 1
- Storage buffer:Phosphate-buffered saline, pH 7.0, no preservative added
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Purified recombinant IL8 (Human).
- Tested applications:ELISA, Neut, WB
- Purification:Protein G purified
- Cat. no.:89360-492
- Supplier No.:GTX18649
Specifications
About this item
IL8, formerly called monocyte derived neutrophil chemotactic factor, belongs to the chemokine alpha family. Human endothelial IL8 is an 8.9 kDa protein containing 77 amino acid residues. IL8 exhibits chemotactic activity in vitro for T cells, basophils and neutrophils and promotes degranulation.
For ELISA: Use at an assay dependent dilution. Can be used as a capture antibody in an ELISA pair with GTX18656 (detection). For Neut: Use at an assay dependent dilution. For WB: Use at a concentration of 0.02 - 0.1 ?g/ml. Predicted molecular weight: 11 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: IL8
Clonality: Monoclonal
Clone: I8-S2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human