To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 1 Alpha
- Clonality:Monoclonal
- Clone:364-3B3-14
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:IL1A
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Antigen synonyms:Hematopoietin-1|IL-1 alpha|pro-interleukin-1-alpha|IL1A|preinterleukin 1 alpha|hematopoietin-1|interleukin-1 alpha
- Storage buffer:Preservative: 0.09% Sodium Azide. Constituents: PBS. pH 7.2
- Storage temperature:–20 °C
- Immunogen:Recombinant human IL-1alpha
- Purification:purified by Protein G chromatography
- Cat. no.:89363-472
- Supplier No.:GTX41494
Specifications
About this item
Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. Each IL acts on a specific, limited group of cells through a receptor specific for that IL. Interleukin 1 (IL1), originally known as lymphocyte activating factor (LAF), activates T cells and lymphocytes, which then proliferate and secrete interleukin 2. IL1 is primarily released from stimulated macrophages and monocytes, but also is released from several other cell types and is thought to play a key role in inflammatory and immune responses. The two closely related agents, interleukin 1 alpha (IL1 alpha) and interleukin 1 beta (IL1 beta) bind to the same cell surface receptor, elicit nearly identical biological responses and share 25% homology in their amino acid sequence. The two closely related agents, interleukin 1 alpha (IL1 alpha) and interleukin 1 beta (IL1 beta) bind to the same cell surface receptor, elicit nearly identical biological responses and share 25% homology in their amino acid sequence.
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 364-3B3-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human