To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Albumin
- Clonality:Monoclonal
- Clone:15C7
- Gene ID:213
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:HSA
- Conjugation:Unconjugated
- ELISA:Yes
- Size:1 mg
- Western blot:Yes
- Antigen synonyms:PRO1341|PRO0883|ANALBA|FDAH|PRO0903
- Storage buffer:Preservative: 0.1% Sodium Azide. Constituents: PBS. pH 7.4
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:8.8 mg/ml
- Immunogen:Human Serum Albumin
- Tested applications:ELISA, WB
- Purification:Protein A purified
- Cat. no.:89367-114
- Supplier No.:GTX40373
Specifications
About this item
Mouse monoclonal [15C7] to HSA
ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 69 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: HSA
Clonality: Monoclonal
Clone: 15C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human