To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:CAMVIR-1
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human papilloma virus (HPV)
- Antigen symbol:HPV 16
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:1 ml
- Storage buffer:Ascites fluid contains less than 0.1% Sodium Azide.
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Immunogen:BALB/C mice were immunized with recombinant HPV-16 L1 protein.
- Tested applications:IHC-P
- Purification:Ascites
- Cat. no.:89353-238
- Supplier No.:GTX73652
Specifications
About this item
Monoclonal Mouse Antibody to Human Papilloma Virus Type 16 ( HPV 16 )
IHC-P: 1:25-1:50 Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with EDTA Buffer (pH 8.0) prior to immunostaining.
Type: Primary
Antigen: HPV 16
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: