To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:HLA class I histocompatibility antigen, alpha chain G
- Clonality:Monoclonal
- Clone:MEM-G/11
- Gene ID:3135
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:HLA-G
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Antigen synonyms:HLA class I histocompatibility antigen|DADB-15K14.8|alpha chain G|HLA-G histocompatibility antigen|b2 microglobulin|MHC-G|HLA class I molecule|HLA G antigen|MHC class I antigen G|class I|G
- Storage buffer:Preservative: Sodium Azide; Constituents: 15mM PBS. pH 7.4
- Storage temperature:Store at 2-8ºC. Do not freeze. Do not use after expiration date stamped on vial label.
- Concentration:1 mg/ml
- Immunogen:Recombinant full length protein (Human) refolded with beta 2 microglobulin and peptide.
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Protein A purified
- Cat. no.:89361-462
- Supplier No.:GTX24570
Specifications
About this item
Mouse monoclonal [MEM - G / 11] to HLA G
ELISA: Use at an assay dependant dilution. FACS: Use at an assay dependant dilution. IP: Use at an assay dependant dilution. Does not cross-block MEM-G/9 (GTX27758 and GTX27904). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human