To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Hepatocyte growth factor
- Clonality:Polyclonal
- Gene ID:3082
- Host:Goat
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:HGF
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Hepatocyte growth factor|HPTA|DFNB39|HGFB|Hepatopoeitin-A|Fibroblast-derived tumor cytotoxic factor|Lung fibroblast-derived mitogen|SF|F-TCF
- Storage buffer:Preservative: NoneConstituents: PBS
- Storage temperature:Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant, human hepatocyte growth factor (rhHGF), expressed in the insect cell line Sf 21.
- Tested applications:ELISA, Neut, WB
- Purification:Protein G purified
- Cat. no.:89354-786
- Supplier No.:GTX10679
Specifications
About this item
Goat polyclonal to HGF
ELISA: Use at a concentration of 0.5 ?g/ml, this detects 0.31ng/well of rhHGF Neut: Use at a concentration of 1 - 16 ?g/ml. WB: Use at a concentration of 1 ?g/ml, this detects rhHGF at 50 ng/lane under both reducing and non-reducing conditions. Predicted molecular weight: 83.1 kDa. Anti-Human HGF was tested for its ability to neutralize the bioactivity of rhHGF in a cell proliferation assay using 4MBr-5 cells, a monkey epithelial cell line responsive to HGF. The ND50 of the antibody is defined as the concentration of antibody resulting in a one-half maximal inhibition of bioactivity of rhHGF, which is present at five times its own EC50 (the concentration of rhHGF producing a one-half maximal bioactivity without antibody). In this bioassay, rhHGF was pre-incubated with various dilutions of the antibody for 1 hour at 37 °C in a 96 well plate. Then, 4MBr-5 cells were added to each well to give a final concentration of 3 x 10^5 cells/ml in 0.1 ml containing 100 ng/ml rhHGF. This was incubated for 48 hours at 37 °C in a 5% CO2 humidified incubator and then pulsed for 4 hours with ^3H-thymidine. Cells were harvested onto glass filters and the ^3H-thymidine incorporation into DNA was measured. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: HGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human