To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glial cell line-derived Neurotrophic Factor
- Clonality:Polyclonal
- Gene ID:2668
- Host:Goat
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:GDNF
- Conjugation:Unconjugated
- ELISA:Yes
- Size:200 μg
- Western blot:Yes
- Antigen synonyms:ATF|GDNF|Astrocyte-derived trophic factor|neurotrophic factor GDNF|Glial cell line-derived neurotrophic factor
- Storage buffer:Constituents: PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant full length human GDNF protein expressed in E. coli.
- Tested applications:I-ELISA
- Purification:Protein G purified
- Cat. no.:89355-470
- Supplier No.:GTX10835
Specifications
About this item
Goat polyclonal to GDNF
I-ELISA: Use at a concentration of 1 ?g/ml detects 0.2 ng/well of rhGDNF. Neut: ND50 = 2.6 ?g/mlWB: Use at a concentration of 2 ?g/ml detects rhGDNF at 1.0 ng/lane under both reducing and non-reducing conditions. Detects a band of approximately 20 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human