To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:EPH Receptor B4
- Clonality:Polyclonal
- Gene ID:13846
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:EPHB4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:25μg
- Antigen synonyms:EPHB4|HTK|MYK1|TYRO11
- Storage buffer:PBS containing 5% trehalose
- Concentration:0.1mg/ml
- Immunogen:Purified recombinant mouse EphB4 extracellular domain expressed in mouse NSO cells.
- Tested applications:Dot Blot, Inhibition
- Purification:Immunogen affinity purified
- Cat. no.:89354-524
- Supplier No.:GTX10618
Specifications
About this item
Goat polyclonal to EphB4
Dot: Use at a concentration of 0.1 - 0.2 ?g/ml. The detection limit for recombinant mouse EphB4 is approximately 5 ng/lane under non-reducing and reducing conditions. ELISA: Use at a concentration of 0.5 - 1.0 ?g/ml, detection limit 1.5 ng/well. Inhib: Use at a concentration of 0.5 - 2 ?g/ml. For neutralization of receptor-ligand interaction, approximately 0.5-2 ?g/ml of the antibody will block 50% of the binding of recombinant mouse ephrin-B2/ (5 ng/ml) to immobilized recombinant mouse EphB4/Fc chimera (100 ?l of a 2 ?g/ml solution coated in each well). The recombinant mouse EphB4 used for the testing of this antibody consists of the extracellular domain of mouse EphB4 (amino acids 1-539) fused by means of a polypeptide linker to the Fc portion of human IgG1 that is histidine-tagged at the C-terminus. N-terminal sequencing indicates that recombinant mouse EphB4/Fc has Leu 16 at the amino terminus. The calculated molecular mass of the reduced protein is approximately 85.2 kDa, but as a result of glycosylation, recombinant EphB4/Fc migrates as an approximately 110 kDa protein under reducing conditions in SDS-PAGE. EphB4 binds to ephrin-B1 and ephrin-B2. Human and mouse EphB4 extracellular domains share approximately 88% homology. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: EPHB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse