To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:EPH Receptor A7
- Clonality:Polyclonal
- Gene ID:2045
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:EPHA7
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:400 μl
- Western blot:Yes
- Antigen synonyms:Receptor protein-tyrosine kinase HEK11|Eph-homology kinase-3|Tyrosine-protein kinase receptor EHK-3|Ephrin type-A receptor 7
- Storage buffer:Phosphate buffered saline containing 0.09% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.25 mg/ml
- Immunogen:Synthetic peptide conjugated to KLH, corresponding to amino acids 911-926 of Human Eph A7.
- Tested applications:ELISA, IHC-F, IHC-P, WB
- Purification:Protein G purified
- Cat. no.:89361-632
- Supplier No.:GTX25411
Specifications
About this item
Rabbit polyclonal to EphA7
For ELISA: Use at a dilution of 1:900 - 1:1,100. For IHC-F and IHC-P: Use at a dilution of 1:50 - 1:100. For WB: Use at a dilution of 1:100 - 1:500. Detects a band of approximately 110 kDa (predicted molecular weight: 119 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: EPHA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human