To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DynActin
- Clonality:Polyclonal
- Gene ID:1639
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:DCTN1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:DAP-150|DP-150|P135
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5 mg/ml
- Immunogen:C-QEQLHQLHSRLIS
- Tested applications:ELISA, IHC, WB
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89358-842
- Supplier No.:GTX11806
Specifications
About this item
The DCTN1 gene encodes the largest subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit interacts with dynein intermediate chain by its domains directly binding to dynein. Alternative splicing of this gene results in at least 2 functionally distinct isoforms: a ubiquitously expressed one and a brain-specific one. Based on its cytogenetic location, this gene is considered as a candidate gene for limb-girdle muscular dystrophy.
Approx 150kDa band observed in cell lline A549 and in Human Testis lysates (calculated MW of 142kDa according to NP_004073.2). Recommended for use at 0.5-2µg/ml. In paraffin embedded Human Cerebellum shows pixulate staining of cytoplasm in neuronal cells. In paraffin embedded Human Testis shows cytoplasm staining in cells of the seminiferous tubules. Recommended concentration, 2-4µg/ml. This antibody is expected to recognise all reported isoforms (NP_004073.2; NP_075408.1; NP_001128512.1; NP_001128513.1).
Type: Primary
Antigen: DCTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse