To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:C-MET
- Conjugation:Unconjugated
- Western blot:Yes
- Modification:phospho-Tyr1349
- Storage buffer:Preservative: 0.05% Sodium Azide; Constituents: 50% glycerol, PBS, 1mg/ml BSA. pH 7.4
- Storage temperature:Store at -80ºC. Upon initial thawing, apportion into working aliquots and store at -80ºC. Avoid repeated freeze-thaw cycles to prevent denaturing the antibody.
- Immunogen:Synthetic phosphopeptide derived from the region of c-Met that contains tyrosine 1349.
- Tested applications:WB
- Purification:Immunogen affinity purified
- Cat. no.:89361-692
- Supplier No.:GTX25669
Specifications
About this item
Binding of scatter factor (SF)/hepatocyte growth factor (HGF) to the c Met receptor tyrosine kinase (RTK) triggers receptor dimerization and phosphorylation on multiple residues within the juxtamembrane, catalytic core and cytoplasmic tail domains, thereby regulating receptor internalization, catalytic activity and multisubstrate docking. c Met contains three tyrosines (Tyrx- x-x-Tyr-Tyr motif) within the activation loop of the catalytic domain, for which phosphorylation of all three (tyrosines 1230, 1234, and 1235) is required for full activation. Activation of the c Met/HGF receptor results in binding and/or phosphorylation of many intracellular signaling proteins including multiple adaptor proteins (e.g., Grb2, Shc, Cbl, Crk, cortactin, paxillin, and GAB1), and a variety of other signal transducers (e.g., PI 3-kinase, FAK, Src, ERK1&2, JNK1&2, PLC gamma, and STAT3). Tyr1349 of c Met is one of the primary docking sites for PI 3-kinase and GAB1. The phosphorylation of tyrosines 1349 and 1356 serves as a priming mechanism for protein binding.
Type: Primary
Antigen: C-MET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: