Anti-CG alpha 4 Mouse Monoclonal Antibody [clone: INN-hFSH-132]
Catalog #: 89357-052
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein Hormones, Alpha Polypeptide
- Clonality:Monoclonal
- Clone:INN-hFSH-132
- Gene ID:1081
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CG alpha 4
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:250 μg
- Western blot:Yes
- Antigen synonyms:CG-ALPHA|FSHA|LHA|GPHa|HCG|TSHA|GPHA1
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Avoid repeated freezing and thawing as this may denature the antibody. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:The recognized epitope comprises amino-acids alpha 15 to alpha 22 of the human sequence.
- Tested applications:Radio Immunoassay
- Purification:Purified IgG prepared by ion exchange chromatography
- Cat. no.:89357-052
- Supplier No.:GTX11232
Specifications
About this item
Mouse monoclonal antibody [INN-hFSH-132] to CG alpha
Type: Primary
Antigen: CG alpha 4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Customers Who Bought This Also Bought