To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of differentiation 8
- Clonality:Monoclonal
- Clone:B334 (H030-1.2)
- Gene ID:925
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Antigen symbol:CD8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:500 μl
- Antigen synonyms:CD8alpha|p32|Leu-2|CD8a|MAL|CD8|CD8b|CD8beta
- Storage buffer:Phosphate-buffered saline, pH 7.2, containing 0.2% BSA and 0.09% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:E-rosette forming cells purified from human PBL
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Protein A affinity purified
- Cat. no.:89353-032
- Supplier No.:GTX72447
Specifications
About this item
Mouse monoclonal [B334 (H030-1.2)] to CD8 T-cell
Immunofluorescence 5-20 ug/mL. Flow cytometry 1-5 ug/10^6 cells. Immunohistochemistry 5-20 ug/mL on frozen sections using avidin-biotin system. The optimal dilution factors should be determined by the individual laboratory. This antibody was not quality controlled in flow cytometry. This antibody can be used on frozen cryostat sections as well as formalin-fixed, paraffin-embedded tissue sections. Prolonged fixation is buffered formalin can destroy the epitope.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: B334 (H030-1.2)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: