To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Transferrin Receptor 1
- Clonality:Monoclonal
- Clone:OX-26
- Gene ID:64678
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Rat
- Antigen symbol:TFR1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:500 μg
- Western blot:Yes
- Antigen synonyms:TRFR|Cluster of Differentiation 71|T9|transferrin receptor|TFR1|IMD46|CD71|p90|TFR|TR
- Storage buffer:Phosphate buffered saline. This product contains sodium.
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles. Should this product contain a precipitate we recommend microcentrifugation before use.
- Concentration:1 mg/ml
- Immunogen:PHA activated rat lymphocytes
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Purified IgG
- Cat. no.:89365-706
- Supplier No.:GTX75149
Specifications
About this item
Mouse monoclonal antibody [OX-26] to CD71
Flow cytometry, WB; Flow Cytometry:Use 10ul of the suggested working dilution to label 10^6 cells in 100ul.
Type: Primary
Antigen: TFR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: