To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD14 Molecule
- Clonality:Polyclonal
- Gene ID:929
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Antigen symbol:CD14
- Conjugation:Unconjugated
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Storage buffer:PBS, no preservatives added
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Amino acids 40-230 of Human CD14.
- Tested applications:WB
- Purification:Affinity purified with antigen
- Cat. no.:89367-224
- Supplier No.:GTX14006
Specifications
About this item
CD14 is a single-copy gene encoding 2 protein forms: a 50- to 55-kD glycosylphosphatidylinositol-anchored membrane protein (mCD14) and a monocyte or liver-derived soluble serum protein (sCD14) that lacks the anchor. Both molecules are critical for lipopolysaccharide (LPS)-dependent signal transduction, and sCD14 confers LPS sensitivity to cells lacking mCD14. Increased sCD14 levels are associated with inflammatory infectious diseases and high mortality in gram-negative shock.
For WB: Use at an assay dependent dilution. Predicted molecular weight: 45 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human