To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Capping protein (actin filament) muscle Z-line, alpha 1
- Clonality:Polyclonal
- Gene ID:829
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Antigen symbol:CAPZA1
- Conjugation:Unconjugated
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:CAPPA1|CAZ1|CAPZ
- Storage buffer:PBS, no preservatives added
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide, corresponding to amino acids 1-286 of CAPZA1.
- Tested applications:WB
- Purification:Affinity purified with antigen
- Cat. no.:89367-430
- Supplier No.:GTX15735
Specifications
About this item
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
For WB: Use at an assay dependent dilution. Predicted molecular weight: 75 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CAPZA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human