To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alkaline Phosphatase, Calf Intestinal
- Clonality:Polyclonal
- Gene ID:280993
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Antigen symbol:ALPI
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:1 mg
- Western blot:Yes
- Storage buffer:0.02M K Phosphate, 0.15M NaCl, 0.01% Sodium Azide, pH7.2
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:10 mg/ml
- Immunogen:Alkaline Phosphatase [Calf intestine]
- Tested applications:Conjugation, Dot Blot
- Purification:IgG fraction
- Cat. no.:89362-552
- Supplier No.:GTX27325
Specifications
About this item
Rabbit polyclonal to calf intestinal Alkaline Phosphatase
Immunoblotting (western or dot blot)ELISA (1:3,000 to 1:15,000)Immunoprecipitation Conjugation and most immunological methods requiring high titer and specificity.
Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine