To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Brain natriuretic peptide
- Clonality:Monoclonal
- Clone:2G9
- Gene ID:4879
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:BNP
- Conjugation:Unconjugated
- ELISA:Yes
- Size:200μg
- Antigen synonyms:brain natriuretic peptide|BNP|NPPB|natriuretic peptide B
- Amino acid number:1 to 32
- Storage buffer:Preservative: 0.1% Sodium Azide; Constituents: PBS
- Sequence:SPKMVQGSGC FGRKMDRISS SSGLGCKVLR RH
- Storage temperature:Store at 2-8 °C.<br />
- Immunogen:Synthetic peptide: SPKMVQGSGC FGRKMDRISS SSGLGCKVLR RH, corresponding to N terminal amino acids 1-32 of Human BNP.
- Tested applications:ELISA
- Purification:Protein A purified
- Cat. no.:89350-904
- Supplier No.:GTX14702
Specifications
About this item
Brain natriuretic peptide (BNP) circulates in blood as a peptide hormone with natriuretic, vasodilatory and renin inhibitory properties. BNP is secreted predominantly by the left ventricular myocytes in response to volume expansion and pressure overload. BNP belongs to a family of structurally similar peptide hormones, which includes atrial natriuretic peptide (ANP), BNP, C-type natriuretic peptide (CNP) and urodilatin. These peptides are characterized by a common 17 amino acid ring structure with a disulfide bond between two cystein residues. This ring structure shows high homology between different natriuretic peptides (eleven of the 17 amino acid residues are homologous in the ring of each of the natriuretic peptides, see fig. 18). BNP is a 32 amino acid peptide with disulfide bond between the cystein residues Cys10 and Cys26. In earlier studies it has been demonstrated that BNP concentration in blood increases with the severity of congestive heart failure. Quantitative measurement of BNP in blood provides an objective indicator of congestive heart failure severity.
ELISA: Use at an assay dependent dilution. EIA: Use at an assay dependent dilution. Can be used for proBNP immunoassay development in combination with anti-NT-proBNP MAbs; GTX13111 Mouse monoclonal [5B6] to proBNP , GTX13115 Mouse monoclonal [15F11] to proBNP and GTX13059 Mouse monoclonal [11D1] to proBNP . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: BNP
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human