To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Brain-derived neurotrophic factor
- Clonality:Monoclonal
- Clone:35928.11
- Gene ID:627
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:BDNF
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Brain-derived neurotrophic factor|ANON2|BDNF|BULN2|Abrineurin
- Storage buffer:Constituents: PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5mg/ml
- Immunogen:Recombinant full length human Brain Derived Neurotrophic Factor protein expressed in Sf21 insect cells.
- Tested applications:I-ELISA
- Purification:Protein A purified
- Cat. no.:89355-454
- Supplier No.:GTX10832
Specifications
About this item
Mouse monoclonal [35928.11] to BDNF
I-ELISA: Use at a concentration of 0.5-1 ?g/ml. WB: Use at a concentration of 1-2 ?g/ml. Predicted molecular weight: 26 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: BDNF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human