To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Anaphase Promoting Complex Subunit 10
- Clonality:Polyclonal
- Gene ID:10393
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:APC10
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:DOC1|APC10
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 and 0.01% (w/v) Sodium Azide
- Storage temperature:Store vial at –20 °C prior to opening. Aliquot contents and freeze at –20 °C or below for extended storage Avoid cycles of freezing and thawing.
- Concentration:1.50 mg/ml
- Immunogen:Synthetic peptide: GADPKQLERTG, corresponding to amino acids 10-20 of Human Apc10.
- Tested applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89367-560
- Supplier No.:GTX23629
Specifications
About this item
Rabbit Polyclonal antibody to APC10
WB: Use at a dilution of 1/500. Detects a band of approximately ~26 kDa (predicted molecular weight: 21 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: APC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human