To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alpha 1 Fetoprotein Receptor
- Clonality:Monoclonal
- Clone:C3
- Gene ID:174
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Dog
- Antigen symbol:AFP
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 μl
- Antigen synonyms:FETA|Alpha-1-Fetoprotein|HPAFP
- Storage buffer:Preservative: Sodium Azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:BALB/C mice were injected with purified human alpha-fetoprotein.
- Tested applications:IHC-P
- Purification:Ascites
- Cat. no.:89351-358
- Supplier No.:GTX17240
Specifications
About this item
This is a major plasma protein produced by the yolk sac and the liver during fetal life. alpha Fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha Fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha Fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. alpha Fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of alpha Fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly.
IHC-P: 1/25 - 1/50. An incubation period of 30 minutes at room temperature is recommended. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog