To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Adenylate Cyclase 3, brain
- Clonality:Polyclonal
- Gene ID:109
- Host:Rabbit
- Isotype:IgG
- Reactivity:Dog
- Antigen symbol:ADCY3
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:AC3
- Storage buffer:Preservative: 0.02% Sodium Azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.75 mg/ml
- Immunogen:The antibodies were generated against a synthetic peptide sequence (unfortunately, the amino acid sequence is considered to be commercially sensitive) unique to PAC3 protein. All peptides were amidated before conjugation.
- Tested applications:WB
- Purification:Immunogen affinity purified
- Cat. no.:89367-398
- Supplier No.:GTX14778
Specifications
About this item
The membrane-bound adenylyl cyclases (ACs) represent one of the major families of effector enzymes for G protein-coupled receptors (GPCRs). Using the high inter-species homology of mammalian AC isoforms, nine Adenylyl cyclase (AC) isoforms, encoded by separate genes, have been identified until today. Human adenylate cyclase genes comprise of 11 to 26 exons, which are distributed over 16 to 430kb. The expression profile of these 9 AC isoforms in a panel of 16 human tissues and in human embryonic kidney (HEK) cells have been demonstrated earlier. The cAMP synthesizing enzymes are found in two forms: cytosolic (soluble) and membrane-bound (particulate). Stimulation of adenylate cyclases produce cAMP form ATP in response to the activation of GPCRs by various hormones, neurotransmitters and other regulatory molecules. cAMP, in subsequent steps down the signal transduction pathway, can stimulate cAMP-dependent protein kinase A (cPKA), and several other target molecules. Activation of cPKA can phosphorylate a broad range of substrates that regulate various metabolic pathways, gene expression, and affect memory functions etc. The stimulation of adenylate cyclases starts with interactions with GPCRs mediated signals initiated by Gs and Gi heterotrimeric G-proteins. The interaction of GPCR agonist (eg. Interaction of isopreternol to beta2 receptors) catalyses the exchange of GDP to GTP that is bound to G proteins. The GTP binding reduces the affinity of Gs'N to other GTP binding proteins and Gs-GTP complex stimulate the adenylate cyclase. In last several years, new members of particulate and soluble adenylate cyclase family have been identified and significant progress is made in understanding of the molecular mechanisms that underlie the regulation of these families of enzymes.
WB: Use at a dilution of 1/500. Detects a band of approximately 119 kDa (predicted molecular weight: 129 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ADCY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog