To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Nestin
- Clonality:Polyclonal
- Gene ID:10763
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:NES
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:C78523|ESTM46|AA166324
- Storage buffer:0.1 ml of affinity purified rabbit antisera in a Tris-citrate/phosphate (pH 7-8) buffer containing 0.1% sodium azide.
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:Recombinant human nestin protein (corresponding to residues 1464-1614).
- Tested applications:WB
- Cat. no.:89352-722
- Supplier No.:GTX30670
Specifications
About this item
Rabbit polyclonal antibody to Nestin - Neural Stem Cell Marker
This antibody may be used in Western blot where a band is seen at ~220-240 kDa. Another band may be seen ~110-115 kDa. Other applications have not been tested. Though this production has not been tested, past productions have been useful for immunohisto/cytochemistry, immunoprecipitation, and FACS analysis of fixed cells.Suggested working dilutions: *Western Blot 1:500 ~ 1:2,000 (ECL)*The investigator should determine the optimal working dilution for a specific application.
Type: Primary
Antigen: NES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human