To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD40 Molecule
- Clonality:Polyclonal
- Gene ID:958
- Host:Rabbit
- Isotype:IgG
- Reactivity:Dog
- Antigen symbol:CD40
- Conjugation:Unconjugated
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:TNF receptor superfamily member 5|TRAP|AI326936|p50|IMD3|GP39|Tnfrsf5|IGM|T-BAM|Bp50|HIGM1
- Storage buffer:PBS containing 50% glycerol and 0.09% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide: CQPVTQEDGKESRISVQERQ conjugated to KLH, corresponding to amino acids 258-277 of Human CD40.
- Tested applications:WB
- Purification:Affinity purified with antigen
- Cat. no.:89359-746
- Supplier No.:GTX13545
Specifications
About this item
CD40 (48 to 50 kDa) is a transmembrane glycoprotein mainly expressed on the surface of B cells and also expressed on monocytes, dendritic cells, and thymic epithelium. CD40 is a member of the tumor necrosis factor (TNF) receptor superfamily, which includes the low affinity nerve growth factor (NGF) receptor and CD95/Fas. CD40 is the receptor for CD40 ligand. CD40 ligand (CD40L, CD154, gp39, and TRAM) belongs to the TNF gene family and is expressed more widely than CD40 predominantly on activated CD4+ T cells. Following interaction with CD40 ligand, CD40 mediates a number of major immunoregulatory functions, central to the control of thymus dependent humoral immunity and may be critical in the development of cell mediated immune responses. Other biological actions include B cell homotypic adhesion, proliferation, immunoglobulin isotype switch, and secretion. Activation of CD40 has also been shown to inhibit the growth of certain B cell lymphomas and to induce the death of transformed cells of mesenchymal or epithelial origin.
- Recommended dilutions: WB: 1/1000. Detects a band of approximately 45 kDa (predicted molecular weight: 34 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: