To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calcitonin-related Polypeptide Alpha
- Clonality:Polyclonal
- Gene ID:796
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CALCA
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:250 μl
- Antigen synonyms:CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
- Storage buffer:Phosphate buffered saline containing 0.1% Sodium Azide (NaN3) 1% Bovine Serum Albumin
- Storage temperature:Store at +4°C. DO NOT FREEZE. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Synthetic human calcitonin
- Tested applications:IHC-P, IHC-Fr
- Purification:Serum
- Cat. no.:89352-342
- Supplier No.:GTX28137
Specifications
About this item
Rabbit polyclonal antibody to Calcitonin
Recommended Starting Dilutions:Immunohistology - Frozen: 1/10 - 1/40Immunohistology - Paraffin: 1/10 - 1/40Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human