To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Phosphodiesterase 11A
- Clonality:Polyclonal
- Gene ID:10846
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:PDE11A
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A|PPNAD2 |PDE11A|cyclic nucleotide phosphodiesterase 11A|FLJ23693|cAMP and cGMP cyclic nucleotide phosphodiesterase 11A|MGC133355|MGC133356|phosphodiesterase 11A
- Storage buffer:'Stabilization buffer' with 0.02% thimerosal or merthiolate.
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.7-0.75 mg/ml
- Immunogen:Synthetic peptide (Human) (N terminal).
- Tested applications:IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89367-360
- Supplier No.:GTX14623
Specifications
About this item
One of the many phosphodiesterases that compartmentalize and hydrolyze cAMP and cGMP into AMP and GMP is Phosphodiesterase type 11. The human PDE11A contains an open reading frame encoding a 490 amino acid protein (55-56 kDa). The PDE11A has an N-terminus GAF domain homologous to other signaling molecule as found in PDE2, PDE5, PDE6, PDE10, which potentially represents an allosteric cGMP and other small signaling molecules. PDE11A hydrolyzes both cGMP and cAMP; it is sensitive to non-selective PDE inhibitor IBMX as well as zaprinast and dipyrimadole inhibitors that are generally selective for cGMP-specific enzymes. PDE11A expression is diverse and is found in high levels in skeletal muscle, prostate, kidney, liver, pituitary and testis.
IP: Use at a dilution of 1/250. WB: Use at a dilution of 1/500. Detects a bands corresponding to at least three PDE11A variants (approximately 57, 65 and 80 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PDE11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human