To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calbindin 2
- Clonality:Polyclonal
- Gene ID:794
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CALB2
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:29kD|CR|CAB29|calretinin|29 kDa Calbindin|Calbindin 2|Calbindin D29K|CAL2
- Storage buffer:Preservative: 0.02% Thimerosal / Merthiolate
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic cyclic peptide (common to all Calretinin proteins) (Human).
- Tested applications:IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89367-384
- Supplier No.:GTX14689
Specifications
About this item
Calretinin is a 29 kDa calcium binding protein that is expressed in central and peripheral nervous system and in many normal and pathological tissues. Calretinin is also considered an important diagnostic aid in the differential diagnosis of cystic and solid ameloblastic tumors. The rat and human calretinin exhibit 98% sequence homology and 91% homology to many other species. Calretinin is generally expressed with two other calcium binding proteins Calbindin D-29 and parvalbumin in CNS and other peripheral tissues. Two calcium-binding proteins, calbindin and calretinin, have been reported to be expressed in abundance in Purkinje cells and other cell types in the cerebellum. The antibodies against calretinin also labels the mossy fibers throughout, and climbing fibers in a distinct group of folia. They also stain several cell types in the molecular layer. There is no appreciable immunostaining of calretinin in cerebellar cortex. In the rat, antibodies against calretinin stain the Lugaro cells, and some granule cells in lobe X; they also give weak staining of all the granule cells in the other lobes. Calretinin antibodies have been used to detect a number of carcinomas by clinicopathological, immunohistochemical techniques including, epithelial malignant mesotheliomas, Ovarian yolk sac tumor, Adenomatoid tumors of the uterus etc.
- IP: Use at a dilution of 1/250. WB: Use at a dilution of 1/1000. Detects a band of approximately 29 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CALB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human