To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calcitonin-related Polypeptide Alpha
- Clonality:Monoclonal
- Clone:SP17
- Gene ID:796
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CALCA
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 μl
- Antigen synonyms:CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
- Storage buffer:This antibody is supplied as tissue culture supernatant containing sodium azide as preservative.
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:Synthetic human calcitonin 1-32 amino acid peptide.
- Tested applications:IHC-P
- Cat. no.:89353-848
- Supplier No.:GTX79412
Specifications
About this item
Monoclonal Rabbit Antibody to Human Calcitonin
We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.
Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: