To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calcitonin-related Polypeptide Alpha
- Clonality:Monoclonal
- Clone:CT10
- Gene ID:796
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CALCA
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 μl
- Antigen synonyms:CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
- Storage buffer:PBS buffer with 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:Synthetic calcitonin.
- Tested applications:IHC-P, IHC-Fr
- Cat. no.:89352-948
- Supplier No.:GTX72079
Specifications
About this item
Mouse Monoclonal to Calcitonin - CT
For IHC: Use at a dilution of 1:40 - 1:80. This antibody can be used on formalin-fixed, paraffin-embedded tissue sections. Prolonged fixation in buffered formalin can destroy the epitope. The antibody may be used at a dilution of 1:40 to 1:80 with AutoProbe III (GTX28620). It is recommended that this product be used on frozen tissue sections or specimens. The optimal conditions should be determined by the researcher.
Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: CT10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human