To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calcitonin-related Polypeptide Alpha
- Clonality:Monoclonal
- Clone:13g11
- Gene ID:796
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CALCA
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
- Storage buffer:Phosphate-buffered saline, pH 7.4, containing 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Concentration:2.3 mg/ml
- Immunogen:Calicitonin protein fragment (purified) (Human) (see figure).
- Tested applications:ELISA, WB
- Purification:Protein A purified
- Cat. no.:89350-920
- Supplier No.:GTX14819
Specifications
About this item
Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. It causes a rapid, but short lived, reduction in serum calcium and phosphate by promoting the incorporation of those ions in the bones. This effect is opposite to that of parathyroid hormone.
For ELISA: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Predicted molecular weight: 13 kDa. Can be paired for ELISA with GTX11494 (conjugate) Mouse monoclonal [42] to Procalcitonin. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 13g11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human