To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Blood Group A Antigen
- Clonality:Monoclonal
- Clone:HE-195
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Antigen symbol:BLOOD GROUP A Antigen
- Conjugation:Unconjugated
- Size:1 mL
- Format:Tissue culture supernatant
- Storage buffer:Preservative: NoneConstituents: Tissue Culture Supernatant
- Storage temperature:–20 °C
- Immunogen:Mixture of erythrocytes of blood group A1 and glycoprotein fraction isolated from the saliva of secretors with blood group A.
- Purification:Tissue culture supernatant
- Cat. no.:89361-264
- Supplier No.:GTX22522
Specifications
About this item
Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Type: Primary
Antigen: BLOOD GROUP A Antigen
Clonality: Monoclonal
Clone: [HE-195]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human