To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:BH3 interacting domain death agonist
- Clonality:Polyclonal
- Gene ID:637
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BID
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:MGC15319|MGC42355|FP497
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-TYVRSLARNGMD
- Tested applications:ELISA, WB
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89366-998
- Supplier No.:GTX89562
Specifications
About this item
Goat polyclonal antibody to Bid
Approx 26+24kDa bands observed in Human fibroblast A431 lysates (calculated MW of 26.8kDa according to NP_932070.1 and 22.0kDa according to NP_001187.1). Recommended concentration: 0.1-0.3µg/ml. This antibody is expected to recognise all three reported isoforms (as represented by NP_932070.1; NP_001187.1; NP_932071.1)
Type: Primary
Antigen: BID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human