To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:IgG
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:1 mg
- Western blot:Yes
- Storage buffer:0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol, pH 8.0 , 10 mg/mL Bovine Serum Albumin (BSA) , 0.1% (w/v) Sodium Azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C. DO NOT FREEZE.
- Concentration:1 mg/ml
- Immunogen:Human IgG whole molecule
- Tested applications:Dot Blot, Immunomicroscopy
- Purification:IgG fraction
- Cat. no.:89352-212
- Supplier No.:GTX26871
Specifications
About this item
AP-conjugated Sheep anti-Human IgG polyclonal antibody
Recommended Starting Dilutions:1:2,000 to 1:8,000 for most applicationsOptimal titers for other applications should be determined by the researcher.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human