Z-Flex 302 Aluminized Heat Resistant Gloves with ZetexPlus Palm, Newtex
Supplier: Newtex Industries
Certificates
About this item
Z-Flex Multi-Layer Aluminization (MLA) fabrics provide superior radiant heat and molten metal splash protection.
- Protection from temperatures up to 1093 °C (2000 °F)
- Cuff protects wrists
- One size fits all
- Made in the USA
The back of the glove is made from Z-Flex Multilayer Aluminized technology, which reflects 95% of radiant heat.
Common applications include; foundries and casting operations, welding and hot works operations, glass and ceramic plants production, power generation, petrochemical and refining, laboratory and testing facilities
The palm of the glove is woven from 100% premium ZetexPlus texturized fiberglass and treated with a proprietary vermiculite coating. Double stitched and reinforced, with a premium wool liner.
Specifications
- Catalog No:
- 89370-994
- 75794-948
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free