About this item
ProSieve™ Protrack™ Dual Color Protein Loading Buffer, the best protection and ability to track protein samples.
- Simple selection - Separation of proteins 5 to 350 kDa in only two range formats
- 20 to 25 minute separation, ProSieve® EX Running Buffer
- 10 minute transfer/Western blot using ProSieve® EX Western Blot Transfer Buffer
- 10 minute staining using ProSieve® EX Safe Stain
- 1 year shelf life
ProSieve™ Protrack™ Dual Color Protein Loading Buffer, protects proteins from degradation during sample preparation with two-color tracking and contains SDS/DTT for complete protein denaturing. 12 month shelf life and storage at 4 degrees. Sustainable and considered non toxic, but should be disposed with proper safety guidelines.