Head, Hickman Still with Side Port, Ace Glass
Catalog #: 89075-190
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Head, Hickman Still with Side Port
- Cat. no.:89075-190
- ST Inner Joint:14/10
- Top ST Joint:[ST] 14/10 (threaded/outer)
- Supplier No.:9576-37
Specifications
About this item
Microscale Hickman still head is used to carry out micro-sample fractional distillations
The top joint is a combination cap, thread and standard taper joint to accommodate and seal other microscale apparatus. Also has an externally threaded side port off reservoir. The O.D. of the top joint has an external thread to facilitate an o-ring and open-top phenolic cap. The internal or I.D. at the top is a ground outer [ST] 14/10 joint. Inner [ST] 14/10 joint at bottom. The inner joint can be secured in reaction vials via “O-Ring-CAP-SEAL” connection. Supplied with cap, o-ring and septa.
Ordering information: Supplied complete with cap for both top and side port, o-ring, and septa. For replacement caps, see 89075-248 (top) or 89075-244 (side port). For replacement o-rings, see 89060-994. For replacement septa, see 89064-076 (top) or 89064-072 (side port).
Customers Who Bought This Also Bought