About this item
Ohaus® VN Series floor platforms and scales offer a unique combination of value, quality, and dependability.
- Weighing
- Parts counting
These floor platforms and scales provide an economical solution to dry weighing needs in industrial and commercial applications
Scales are supplied complete with durable T31P Indicators. They offer selectable weight displays in kg or lb. measurements. Standard features include counting mode, ball-in-cup leveling feet, steel eye bolts for easy lifting and positioning, and a bi-directional interface for printing or connection to a PC.
Additional standard features include, ABS Indicator, a black painted steel platform, raised embossed treads, fixed top plate (5mm thick), four IP67 NTEP alloy steel load cells, and an IP65 stainless steel junction box with side access.
The high-quality construction is designed to minimize distortion under heavy loads, and ensures years of dependable use. NTEP certification ensures consistent weighing performance. Scales are available with ramps and mounting feet.
Weight: 2268 kg (5000 lbs.)